Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA567390
Description
Immunogen sequence: SKSSVKVMLSK TMDKENRPEA VKSCEALAQK IEEFLEAHST DEFKDVSATE PQTAHSMTNR FNAMLKVFEN Q Highest antigen sequence identity to the following orthologs - mouse 40%, rat 42%.
May play a role in tumorigenesis.
Specifications
CCDC7 | |
Polyclonal | |
Unconjugated | |
CCDC7 | |
BIOT2; BioT2-A; BioT2-B; BioT2-C; C10orf68; CCDC7; coiled-coil domain containing 7; Coiled-coil domain-containing protein 7; coiled-coil domain-containing protein 7; uncharacterized protein C10orf68; Protein BIOT2; RP11-479G22.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79741 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96M83 | |
CCDC7 | |
Recombinant Human CCDC7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction