Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC71 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561543
Description
Immunogen sequence: MSVVVQHVEE KAVHSWSRIS TAGKKALEEA LLVFNPMSQD LSATEAQLVA FLQGLRDDGF QPTILRSGDV YGYSSCTANP PSQTKLQARA PNPTAT Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

Specifications
CCDC71 | |
Polyclonal | |
Unconjugated | |
Ccdc71 | |
2600016J21Rik; CCDC71; coiled-coil domain containing 71; coiled-coil domain-containing protein 71 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64925 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IV32 | |
Ccdc71 | |
Recombinant protein corresponding to Human CCDC71. Recombinant protein control fragment (Product #RP-100241). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction