Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC74A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570393 Supplier Invitrogen™ Supplier No. PA570393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 86% Synthetic peptide located within the following region: FPKVSTK SLSKKCLSPP VAERAILPAL KQTPKNNFAE RQKRLQAMQK RRL.

CCDC74A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC74A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCDC74A
Gene Accession No. Q96AQ1
Gene Alias 2310015A05Rik; CCDC74A; coiled-coil domain containing 74A; coiled-coil domain-containing protein 74A; coiled-coil domain-containing protein 74B
Gene Symbols CCDC74A
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human CCDC74A (aa 324-373).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90557
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.