Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC75 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306106F
Description
CCDC75 Polyclonal specifically detects CCDC75 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
CCDC75 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
CENP-Y, coiled-coil domain containing 75 | |
Produced in rabbits immunized with E. coli-derived Human CCDC75 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
253635 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction