Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC76 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232177APC
Description
CCDC76 Polyclonal antibody specifically detects CCDC76 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
CCDC76 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
coiled-coil domain containing 76, Coiled-coil domain-containing protein 76, EC 2.1.1.-, FLJ10287, FLJ11219, tRNA [Gm4] methyltransferase, tRNA guanosine-2'-O-methyltransferase TRM13 homolog | |
The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human Coiled-Coil Domain Containing 76 using the numbering given in entry NP_061956.2 | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
54482 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction