Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC78 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544001
Description
Peptide sequence: QELRHKAQVP GHSDDHRFQV QPKNTMDPEN EQHRLGSGVS VQPPSSGERA Sequence homology: Human: 93%.
The exact function of CCDC78 remains unknown.
Specifications
| CCDC78 | |
| Polyclonal | |
| Unconjugated | |
| CCDC78 | |
| C16orf25; CCDC78; CNM4; coiled-coil domain containing 78; coiled-coil domain-containing protein 78; hsCCDC78; JFP10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 124093 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A2IDD5 | |
| CCDC78 | |
| Synthetic peptide directed towards the middle region of human CCDC78. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction