Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC8 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP150022JF646
Description
CCDC8 Polyclonal antibody specifically detects CCDC8 in Human samples. It is validated for Western BlotSpecifications
CCDC8 | |
Polyclonal | |
Western Blot | |
coiled-coil domain containing 8, coiled-coil domain-containing protein 8, DKFZp564K0322 | |
The immunogen for this product maps to a region between residue 488 and 538 of human Coiled-Coil Domain Containing 8 using the numbering given in entry NP_114429.2 (GeneID 83987). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
83987 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction