Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559811
Description
Immunogen sequence: DNHREEAADN QRAEAPADQG SEVTDNQREE AVHDQRERAP AVQGADNQRA QARAGQRAEA AHNQRAGAPG IQEAEVSAAQ G Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 43%.
The microphthalmia transcription factor/transcription factor E (MITF-TFE) family of basic helix-loop-helix leucine zipper (bHLH-Zip) transcription factors includes four family members: MITF, TFE3, TFEB and TFEC. The TEF3 protein encoded by this gene activates transcription through binding to the muE3 motif of the immunoglobulin heavy-chain enhancer. The TFEC protein forms heterodimers with the TEF3 protein and inhibits TFE3-dependent transcription activation. The TEF3 protein interacts with transcription regulators such as E2F3, SMAD3, and LEF-1, and is involved in TGF-beta-induced transcription, playing important roles in cell growth, proliferation, and osteoclast and macrophage differentiation. The TFE3 protein also activates hepatic IRS-2 gene, and induces hexokinase II (HK2) and insulin-induced gene 1 (INSIG1); it participates in insulin signaling and could be a therapeutic target for diabetes. This gene is also involved in chromosomal translocations, resulting in different fusion gene products in papillary renal cell carcinomas and alveolar soft part sarcomas, such as PRCC-TFE3, RCC17-TFE3, PSF-TFE3, NonO (p54nrb)-TFE3 and ASPL-TFE3.
Specifications
CCDC8 | |
Polyclonal | |
Unconjugated | |
CCDC8 | |
3M3; CCDC8; coiled-coil domain containing 8; coiled-coil domain containing protein 8; coiled-coil domain-containing protein 8; Coiled-coil domain-containing protein 8 homolog; ENSMUSG00000041117; p90; PPP1R20; probable Coiled-coil domain-containing protein 8; protein phosphatase 1, regulatory subunit 20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
83987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H0W5 | |
CCDC8 | |
Recombinant protein corresponding to Human CCDC8. Recombinant protein control fragment (Product #RP-99167). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction