Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC80 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551695 Supplier Invitrogen™ Supplier No. PA551695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMSLLKDDVY CELAERHIQQ IVLFHQAGEE GGKVRRITSE GQILEQPLDP SLIPKLMSFL KLEKGKFGMV LLKKTLQVEE RYPYPVRLEA MYEVIDQGPI RRIEKIRQKG FVQKCKASGV EGQVVAEGND GGG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

CCDC80 (Coiled-Coil Domain Containing 80) is a Protein Coding gene. Diseases associated with CCDC80 include Herpes Zoster Oticus and Peripheral Nervous System Disease. Gene Ontology (GO) annotations related to this gene include heparin binding and glycosaminoglycan binding. Also, it is an estrogen regulated gene; overexpression correlates with mammary carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC80
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC80
Gene Accession No. Q76M96
Gene Alias 2610001E17Rik; Ccdc80; CL2; coiled-coil domain containing 80; coiled-coil domain-containing protein 80; coiled-coil domain-containing protein 80 pseudogene; down-regulated by oncogenes 1 protein; Down-regulated by oncogenes protein 1; DRO1; HBE245; LOW QUALITY PROTEIN: coiled-coil domain-containing protein 80; nuclear envelope protein okuribin; okuribin; Ssg1; SSG-1; steroid sensitive gene 1; steroid sensitive gene-1 protein; steroid-sensitive gene 1 protein; steroid-sensitive protein 1; up-regulated in BRS-3 deficient mouse; Up-regulated in BRS-3 deficient mouse homolog; Urb
Gene Symbols CCDC80
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC80. Recombinant protein control fragment (Product #RP-101003).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151887
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.