Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC80 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988190
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8190 Supplier Bioss Supplier No. BS7992R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC80 (Coiled-Coil Domain Containing 80) is a Protein Coding gene. Diseases associated with CCDC80 include Herpes Zoster Oticus and Peripheral Nervous System Disease. Gene Ontology (GO) annotations related to this gene include heparin binding and glycosaminoglycan binding. Also, it is an estrogen regulated gene; overexpression correlates with mammary carcinogenesis.

Specifications

Antigen CCDC80
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC80
Gene Accession No. Q76M96
Gene Alias 2610001E17Rik; BOS_752; CCDC80; CL2; coiled-coil domain containing 80; Coiled-coil domain-containing protein 80; Down-regulated by oncogenes 1 protein; Down-regulated by oncogenes protein 1; DRO1; HBE245; LOW QUALITY PROTEIN: coiled-coil domain-containing protein 80; nuclear envelope protein okuribin; okuribin; Ssg1; SSG-1; steroid sensitive gene 1; steroid sensitive gene-1 protein; Steroid-sensitive gene 1 protein; steroid-sensitive protein 1; Up-regulated in BRS-3 deficient mouse; Up-regulated in BRS-3 deficient mouse homolog; Urb
Gene Symbols CCDC80
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC80.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151887
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.