Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC82 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128743DL594
Description
CCDC82 Polyclonal antibody specifically detects CCDC82 in Human samples. It is validated for Western BlotSpecifications
CCDC82 | |
Polyclonal | |
Western Blot | |
coiled-coil domain containing 82, coiled-coil domain-containing protein 82, FLJ23518, HSPC048 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human coiled-coil domain containing 82 using the numbering given in entry NP_079001.2 (GeneID 79780). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79780 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction