Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC82 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546237
Description
Peptide sequence: RVDWRRTKRS SISQLLDSDE ELDSEEFDSD EELDSDESFE NDEELDSNKG Sequence homology: Human: 100%; Yeast: 83%.
The function of this protein remains unknown.
Specifications
| CCDC82 | |
| Polyclonal | |
| Unconjugated | |
| CCDC82 | |
| 2310043N13Rik; CCDC82; coiled-coil domain containing 82; coiled-coil domain-containing protein 82; FLJ23518; HSPC048; HT025 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79780 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N4S0 | |
| CCDC82 | |
| Synthetic peptide directed towards the N-terminal of human CCDC82 (aa 23-72). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction