Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC82 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546778 Supplier Invitrogen™ Supplier No. PA546778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SSDEVDEEEE EDNYESDEDG DDYIIDDFVV QDEEGDEENK NQQGEKLTTS.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC82
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCDC82
Gene Accession No. Q8N4S0
Gene Alias 2310043N13Rik; CCDC82; coiled-coil domain containing 82; coiled-coil domain-containing protein 82; FLJ23518; HSPC048; HT025
Gene Symbols CCDC82
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CCDC82 (aa 271-320).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79780
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.