Learn More
Invitrogen™ CCDC83 Monoclonal Antibody (3E3)

Description
Peptide Sequence: SSDESTILHL SHENSIEDLQ YVKIDKEENS GTEFGDTDMK YLLYEDEKDF KDYVNLGPLG VKLMSVESKK MPIHFQEKEI PVKLYKDVRS PESHITYKMM KSFL.
Specifications
Specifications
| Antigen | CCDC83 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3E3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CCDC83 |
| Gene Accession No. | Q8IWF9 |
| Gene Alias | 4930549K11Rik; 4930554C01Rik; 4932423M01Rik; CCDC83; coiled-coil domain containing 83; coiled-coil domain-containing protein 83; CT148; HSD9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.