Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC83 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109486 Supplier Invitrogen™ Supplier No. PA5109486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MENSGKANKK DTHDGPPKEI KLPTSEALLD YQCQIKEDAV EQFMFQIKTL RKKNQKYHER NSRLKEEQIW HIRHLLKELS EEKAEGLPVV TREDVEEAMK EKWKFERDQE KNLRDMRMQI SNAEKLFLEK LSEKEYWEEY KNVGSERHAK LITSLQNDIN TVKENAEKMS EHYKITLEDT RKKIIKETLL QLDQKKEWAT QNAVKLIDKG SYLEIWENDW LKKEVAIHRK EVEELKNAIH ELEAENLVLI DQLSNCRLVD LKIPRRLYLT QAAGLEVPPE EMSLELPETH IEEKSELQPT.

CCDC83 (Coiled-Coil Domain Containing 83) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC83
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CCDC83
Gene Accession No. Q8IWF9
Gene Alias 4930549K11Rik; 4930554C01Rik; 4932423M01Rik; CCDC83; coiled-coil domain containing 83; coiled-coil domain-containing protein 83; CT148; HSD9
Gene Symbols CCDC83
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CCDC83 (NP_001273088.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220047
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.