Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC84 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558976 Supplier Invitrogen™ Supplier No. PA558976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEKFLVTPQD YARFKKSMVK GLDSYEEKED KVIKEMAAQI REVEQSRQEV VRSVLEPQAV PDPEEGSSAP RSWKGMNSQV ASSLQQPSNL DLPP Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

This gene encodes a protein thought to contain a coiled coil motif. No function has been determined for the encoded protein. A pseudogene of this gene is located on chromosome 20. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Aug 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC84
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC84
Gene Accession No. Q86UT8
Gene Alias CCDC84; coiled-coil domain containing 84; coiled-coil domain-containing protein 84; D630044F24Rik; DLNB14; Gm1114
Gene Symbols CCDC84
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC84. Recombinant protein control fragment (Product #RP-97530).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338657
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.