Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC84 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548182
Description
This gene encodes a protein thought to contain a coiled coil motif. No function has been determined for the encoded protein. A pseudogene of this gene is located on chromosome 20. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Aug 2013]
Specifications
CCDC84 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q86UT8, Q4VA36 | |
CCDC84 | |
KLH conjugated synthetic peptide between 34-62 amino acids from the N-terminal region of human CCDC84. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CCDC84 | |
Coiled-coil domain-containing protein 84, CCDC84 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
338657, 382073 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction