Learn More
Invitrogen™ CCDC85A Polyclonal Antibody

Description
Immunogen sequence: DDDRQKGKRV SREWQRLGRY TAGVMHKEVA LYLQKLKDLE VKQEEVVKEN MELKELCVLL DEEKGAGCAG SRCSI Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
Specifications
Specifications
| Antigen | CCDC85A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CCDC85A |
| Gene Accession No. | Q96PX6 |
| Gene Alias | Ccdc85a; coiled-coil domain containing 85A; coiled-coil domain-containing protein 85A; KIAA1912; RGD1311700; zgc:175167 |
| Gene Symbols | CCDC85A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.