Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC85A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570880
Description
This target displays homology in the following species: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
CCDC85A is a protein coding gene.
Specifications
CCDC85A | |
Polyclonal | |
Unconjugated | |
CCDC85A | |
Ccdc85a; coiled-coil domain containing 85A; coiled-coil domain-containing protein 85A; KIAA1912; RGD1311700; zgc:175167 | |
Rabbit | |
Affinity chromatography | |
RUO | |
114800 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96PX6 | |
CCDC85A | |
synthetic peptide directed towards the N-terminal region of human CCDC85A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction