Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC85C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563559
Description
Immunogen sequence: PSAGYSPAGQ KPEAVVHAMK VLEVHENLDR QLQDSCEEDL SEKEKAIVRE MCNVVWR Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Specifications
CCDC85C | |
Polyclonal | |
Unconjugated | |
CCDC85C | |
Ccdc85c; coiled-coil domain containing 85C; coiled-coil domain-containing protein 85C; EG668158; Gm265; Gm9010 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
317762 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NKD9 | |
CCDC85C | |
Recombinant protein corresponding to Human CCDC85C. Recombinant protein control fragment (Product #RP-100320). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction