Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC86 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559433
Description
Immunogen sequence: LEEEKERRRQ EKKQRRAENL KRRLENERKA EVVQVIRNPA KLKRAKKKQL RSIEKRDTLA LLQKQPPQQP AAKI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
Coiled-coil domain-containing protein 86 (CCDC86) was identified as an immediate-early cytokine-responsive gene induced by interleukin 3 (IL-3) in a hematopoietic cell line. CCDC86 contains a coiled-coil domain in the C-terminus.
Specifications
CCDC86 | |
Polyclonal | |
Unconjugated | |
CCDC86 | |
4933411H20Rik; 6720480F16Rik; CCDC86; coiled-coil domain containing 86; coiled-coil domain-containing protein 86; CYCLON; cytokine-induced protein with coiled-coil domain; D19Ertd678e; hypothetical protein LOC540913; mCyclon; MGC2574; MNCb-4327 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79080 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6F5 | |
CCDC86 | |
Recombinant protein corresponding to Human CCDC86. Recombinant protein control fragment (Product #RP-97315). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction