Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC86 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559617 Supplier Invitrogen™ Supplier No. PA559617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQQGAGLESP QGQPEPGAAS PQRQQDLHLE SPQRQPEYSP ESPRCQPKPS EEAPKCSQDQ GVLASELAQN KEELTPGAPQ HQLPPV Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 42%.

Coiled-coil domain-containing protein 86 (CCDC86) was identified as an immediate-early cytokine-responsive gene induced by interleukin 3 (IL-3) in a hematopoietic cell line. CCDC86 contains a coiled-coil domain in the C-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC86
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC86
Gene Accession No. Q9H6F5
Gene Alias 4933411H20Rik; 6720480F16Rik; CCDC86; coiled-coil domain containing 86; coiled-coil domain-containing protein 86; CYCLON; cytokine-induced protein with coiled-coil domain; D19Ertd678e; hypothetical protein LOC540913; mCyclon; MGC2574; MNCb-4327
Gene Symbols CCDC86
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC86. Recombinant protein control fragment (Product #RP-97314).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79080
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.