Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC87 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544709 Supplier Invitrogen™ Supplier No. PA544709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MMEPPKPEPE LQRFYHRLLR PLSLFPTRTT SPEPQKRPPQ EGRILQSFPL Sequence homology: Human: 100%.

Plays a role in spermatogenesis, where it is important for normal sperm head morphology. Also required for the acrosome reaction and thus normal male fertility. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC87
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CCDC87
Gene Accession No. Q9NVE4
Gene Alias 4931419P11Rik; Ccdc87; coiled-coil domain containing 87; coiled-coil domain-containing protein 87
Gene Symbols CCDC87
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CCDC87 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55231
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.