Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC88B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988308
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8308 Supplier Bioss Supplier No. BS8141R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen CCDC88B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC88B
Gene Accession No. A6NC98
Gene Alias 78 kDa glucose-regulated protein [GRP78]-interacting protein induced by ER stress; Brain leucine zipper domain-containing protein; brain leucine zipper protein; BRLZ; CCDC88; CCDC88B; coiled-coil domain containing 88; coiled-coil domain containing 88B; Coiled-coil domain-containing protein 88B; Gipie; GRP78-interacting protein induced by ER stress; HkRP3; Hook-related protein 3
Gene Symbols CCDC88B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC88B/BRLZ.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283234
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.