Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC89 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549671
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549671 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549671 Supplier Invitrogen™ Supplier No. PA549671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC89 Polyclonal Antibody for Western Blot

CCDC89 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC89
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene zgc:172182
Gene Accession No. A8WG43
Gene Alias 1700019B01Rik; AU022100; bc8 orange-interacting protein; bc8 orange-interacting protein {ECO:0000303; BOIP; ccdc89; ccdc89 {ECO:0000312; coiled-coil domain containing 89; coiled-coil domain-containing protein 89; coiled-coil domain-containing protein 89 {ECO:0000312; ECO:0000312; EMBL:AAI67063.1, ECO:0000312; EMBL:AAI670631; PubMed:14648848}; RGD:1305335}; RGD1305335; zgc:172182
Gene Symbols zgc:172182
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 265-293 amino acids from the C-terminal region of DANRE ccdc89.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100136847
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.