Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ccdc89 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565274
Description
Immunogen sequence: PEERLEKQNEK LNNQEEETEF KELDGLREAL ANLRGLSEEE RSEKAMLRSR IEEQSQLICI LKRRSDEALE RCQILELLNA ELEEKMMQE Highest antigen sequence identity to the following orthologs - mouse 83%, rat 81%.
CCDC89 is a protein coding gene.
Specifications
Ccdc89 | |
Polyclonal | |
Unconjugated | |
CCDC89 | |
1700019B01Rik; AU022100; bc8 orange-interacting protein; bc8 orange-interacting protein {ECO:0000303; BOIP; ccdc89; ccdc89 {ECO:0000312; coiled-coil domain containing 89; coiled-coil domain-containing protein 89; coiled-coil domain-containing protein 89 {ECO:0000312; ECO:0000312; EMBL:AAI67063.1, ECO:0000312; EMBL:AAI670631; PubMed:14648848}; RGD:1305335}; RGD1305335; zgc:172182 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
220388 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N998 | |
CCDC89 | |
Recombinant Human Ccdc89. Recombinant protein control fragment (Product #RP-104722). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction