Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561054
Description
Immunogen sequence: IEEDRKKAEL EGVAVTAPRK GRSVEKENVA VESEKNLGPS RRSPGTPRPP GASKGGRTPP QQGGRAGMGR ASRSWE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
Coiled-coil domain-containing protein 9 (CCDC9) contains one coiled-coil domain.
Specifications
CCDC9 | |
Polyclonal | |
Unconjugated | |
CCDC9 | |
2600011L02Rik; Ccdc9; coiled-coil domain containing 9; coiled-coil domain-containing protein 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26093 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3X0 | |
CCDC9 | |
Recombinant protein corresponding to Human CCDC9. Recombinant protein control fragment (Product #RP-100047). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction