Learn More
Invitrogen™ CCDC90A Polyclonal Antibody

Description
Peptide sequence: ELHQLKQQVM DEVIKVRTDT KLDFNLEKSR VKELYSLNEK KLLELRTEIV Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rat: 93%.
Specifications
Specifications
| Antigen | CCDC90A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | MCUR1 |
| Gene Accession No. | Q96AQ8 |
| Gene Alias | 6230416A05Rik; AU015498; AV136929; AW554392; C6orf79; C88263; Ccdc90a; coiled-coil domain containing 90A; coiled-coil domain-containing protein 90A, mitochondrial; FMP32; formin-like protein 16; MCU regulator 1; MCUR1; MCURI1; Mitochondrial calcium uniporter regulator 1; RGD1307673 |
| Gene Symbols | MCUR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.