Learn More
Invitrogen™ CCDC90A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595628
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cellhuman MDA-MB-231 whole cell, human HL-60 whole cell, human MDA-MB-453 whole cell, human A431 whole cell, human Caco-2 whole cell, rat spleen tissue, mouse lung tissue, mouse Ana-1 whole cell. IHC: human oesophagus squama cancer tissue, human ovary cancer tissue, human lung cancer tissue, human placenta tissue, human tonsil tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Mcur1 encodes a protein that is a key regulator of mitochondrial calcium uniporter (MCU) required for calcium entry into mitochondrion. Mcur1 plays a direct role in uniporter-mediated calcium uptake via a direct interaction with MCU (mitochondrial calcium uniporter). Mcur1 may be involved in the assembly of the membrane components of the uniporter complex.
Specifications
CCDC90A | |
Polyclonal | |
Unconjugated | |
MCUR1 | |
6230416A05Rik; AU015498; AV136929; AW554392; C6orf79; C88263; Ccdc90a; coiled-coil domain containing 90A; coiled-coil domain-containing protein 90A, mitochondrial; FMP32; formin-like protein 16; MCU regulator 1; MCUR1; MCURI1; Mitochondrial calcium uniporter regulator 1; RGD1307673 | |
Rabbit | |
Affinity chromatography | |
RUO | |
291034, 63933, 76137 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
Q96AQ8, Q9CXD6 | |
MCUR1 | |
A synthetic peptide corresponding to a sequence of human MCUR1 (ATQQAEIIVSALVKILEANMDIVYKDMVTKMQQE). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.