Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC90A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987601
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7601 Supplier Bioss Supplier No. BS6924R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mcur1 encodes a protein that is a key regulator of mitochondrial calcium uniporter (MCU) required for calcium entry into mitochondrion. Mcur1 plays a direct role in uniporter-mediated calcium uptake via a direct interaction with MCU (mitochondrial calcium uniporter). Mcur1 may be involved in the assembly of the membrane components of the uniporter complex.

Specifications

Antigen CCDC90A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MCUR1
Gene Accession No. Q96AQ8
Gene Alias 6230416A05Rik; AU015498; AV136929; AW554392; BOS_21897; C6orf79; C88263; CCDC90A; coiled-coil domain containing 90A; Coiled-coil domain-containing protein 90A, mitochondrial; FMP32; MCU regulator 1; Mcur1; Mitochondrial calcium uniporter regulator 1; RGD1307673
Gene Symbols MCUR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC90A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63933
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.