Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC90B Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305879AF750
Description
CCDC90B Polyclonal specifically detects CCDC90B in Human samples. It is validated for Western Blot.Specifications
CCDC90B | |
Polyclonal | |
Western Blot | |
coiled-coil domain containing 90B, mitochondrial | |
Produced in rabbits immunized with E. coli-derived Human CCDC90B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
60492 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction