Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC90B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552795 Supplier Invitrogen™ Supplier No. PA552795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHLDAIRKDM VILEKSEFAN LRAENEKMKI ELDQVKQQLM HETSRIRADN KLDINLERSR VTDMFTDQEK QLMETTTEFT KKDTQTKSII SETSNKIDAE IASLKTLMES NKLE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

CCDC90B (Coiled-Coil Domain-Containing 90B), also known as MCUR1 (Mitochondrial Calcium Uniporter Regulator 1), is a gene located on chromosome 16p13.3 that encodes a protein involved in mitochondrial function. MCUR1 is a critical component of the mitochondrial calcium uniporter complex, which plays a vital role in regulating mitochondrial calcium uptake. Calcium uptake by mitochondria is essential for cellular metabolism, energy production, and apoptosis. MCUR1 interacts with other components of the uniporter complex to modulate the entry of calcium into the mitochondrial matrix, thereby influencing mitochondrial bioenergetics and reactive oxygen species (ROS) generation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC90B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC90B
Gene Accession No. Q9GZT6
Gene Alias 2310015N07Rik; C86558; CCDC90B; coiled-coil domain containing 90B; coiled-coil domain-containing protein 90B, mitochondrial; CUA003; MDS011; MDS025; RGD1308637
Gene Symbols CCDC90B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC90B. Recombinant protein control fragment (Product #RP-90270).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60492
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.