Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC90B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117844 Supplier Invitrogen™ Supplier No. PA5117844
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specificity: Human CCDC90B.

CCDC90B (Coiled-Coil Domain-Containing 90B), also known as MCUR1 (Mitochondrial Calcium Uniporter Regulator 1), is a gene located on chromosome 16p13.3 that encodes a protein involved in mitochondrial function. MCUR1 is a critical component of the mitochondrial calcium uniporter complex, which plays a vital role in regulating mitochondrial calcium uptake. Calcium uptake by mitochondria is essential for cellular metabolism, energy production, and apoptosis. MCUR1 interacts with other components of the uniporter complex to modulate the entry of calcium into the mitochondrial matrix, thereby influencing mitochondrial bioenergetics and reactive oxygen species (ROS) generation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC90B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CCDC90B
Gene Accession No. Q9GZT6
Gene Alias 2310015N07Rik; C86558; CCDC90B; coiled-coil domain containing 90B; coiled-coil domain-containing protein 90B, mitochondrial; CUA003; MDS011; MDS025; RGD1308637
Gene Symbols CCDC90B
Host Species Rabbit
Immunogen E. coli-derived Human CCDC90B fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60492
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.