Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC90B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531579
Description
Recommended positive controls: MCF-7, mouse heart. Predicted reactivity: Mouse (91%), Rat (88%), Rhesus Monkey (97%), Chimpanzee (99%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
CCDC90B | |
Polyclonal | |
Unconjugated | |
CCDC90B | |
2310015N07Rik; C86558; CCDC90B; coiled-coil domain containing 90B; coiled-coil domain-containing protein 90B, mitochondrial; CUA003; MDS011; MDS025; RGD1308637 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60492, 66365 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8C3X2, Q9GZT6 | |
CCDC90B | |
Recombinant fragment corresponding to a region within amino acids 1 and 195 of Human CCDC90B. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction