Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC91 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558280 Supplier Invitrogen™ Supplier No. PA558280
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDDDDFGGFE AAETFDGGSG ETQTTSPAIP WAAFPAVSGV HLSPSSPEIV LDRDHSSSIG CLSSDAIISS PENTHAANSI VSQTIPKAQ Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.

CCDC91 (Coiled-Coil Domain-Containing 91) is a gene located on chromosome 12q24.31 that encodes a protein involved in vesicle-mediated transport and Golgi function. The CCDC91 protein contains coiled-coil domains, which facilitate protein-protein interactions necessary for its role in the secretory pathway. Specifically, CCDC91 is a component of the Golgi-associated retrograde protein (GARP) complex, which is essential for the recycling of transport vesicles from endosomes to the trans-Golgi network (TGN). Research has revealed that CCDC91 is crucial for maintaining Golgi structure and function and is involved in the sorting and trafficking of proteins and lipids within the cell. The protein's interactions with other components of the vesicular transport machinery enable the efficient movement of cargo between organelles, which is critical for various cellular processes, including secretion, endocytosis, and lysosomal degradation. Mutations or dysregulation of CCDC91 can disrupt these processes, potentially leading to diseases associated with vesicle trafficking defects, such as certain neurodegenerative disorders and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC91
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC91
Gene Accession No. Q7Z6B0
Gene Alias 1700086G08Rik; 1810060J02Rik; AU045683; AW555238; Ccdc91; coiled-coil domain containing 91; coiled-coil domain-containing protein 91; GGA binding partner; GGA-binding partner; Ggabp; HSD8; p56; p56 accessory protein
Gene Symbols CCDC91
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC91. Recombinant protein control fragment (Product #RP-97723).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55297
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.