Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC93 Antibody (OTI6E11), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271963AF405
Description
CCDC93 Monoclonal specifically detects CCDC93 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CCDC93 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CCDC93 | |
Recombinant protein fragment corresponding to amino acids 293-631 of human CCDC93 (NP_061917) produced in E.coli. | |
0.1 mL | |
54520.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI6E11 | |
Western Blot, Immunohistochemistry | |
coiled-coil domain containing 93, coiled-coil domain-containing protein 93, FLJ10996, FLJ25197, MGC13033 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction