Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC93 Antibody (OTI6E11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245588
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
CCDC93 Monoclonal specifically detects CCDC93 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CCDC93 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
coiled-coil domain containing 93, coiled-coil domain-containing protein 93, FLJ10996, FLJ25197, MGC13033 | |
Mouse | |
73 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Western Blot, Immunohistochemistry | |
OTI6E11 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
CCDC93 | |
Human recombinant protein fragment corresponding to amino acids 293-631 of human CCDC93 (NP_061917) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
54520 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction