Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC93 Monoclonal Antibody (OTI6E11)
GREENER_CHOICE

Catalog No. PIMA526464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526464 Supplier Invitrogen™ Supplier No. MA526464
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCDC93 Monoclonal Antibody for Western Blot, IHC (P)

CCDC93 is involved in copper-dependent ATP7A trafficking between the trans-Golgi network and vesicles in the cell periphery; the function is proposed to depend on its association within the CCC complex and cooperation with the WASH complex on early endosomes and is dependent on its interaction with WASHC2C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC93
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCDC93
Gene Accession No. Q567U6
Gene Alias 4633402D15Rik; 9230102M16Rik; CCDC93; coiled-coil domain containing 93; coiled-coil domain-containing protein 93
Gene Symbols CCDC93
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 293-631 of CCDC93 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 476119, 54520
Target Species Human, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.