Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC94 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB276078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB276078 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB276078 Supplier Novus Biologicals Supplier No. NBP318502JF549
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC94 Polyclonal antibody specifically detects CCDC94 in Human samples. It is validated for Western Blot

Specifications

Antigen CCDC94
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias coiled-coil domain containing 94, coiled-coil domain-containing protein 94, FLJ10374
Host Species Rabbit
Immunogen Recognizes region between residues Between between 273 and 323..
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55702
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.