Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC94 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560435 Supplier Invitrogen™ Supplier No. PA560435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITFKTDPENT DYTMEHGATR NFQAEKLLEE EEKRVQKERE DEELNNPMKV LENRTKDSKL EMEVLENLQE LK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

Part of the spliceosome which catalyzes two sequential transesterification reactions, first the excision of the non-coding intron from pre-mRNA and then the ligation of the coding exons to form the mature mRNA (PubMed:29301961). Plays a role in stabilizing the structure of the spliceosome catalytic core and docking of the branch helix into the active site, producing 5'-exon and lariat intron-3'-intermediates. May protect cells from TP53-dependent apoptosis upon dsDNA break damage through association with PRP19-CD5L complex (PubMed:22952453). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC94
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC94
Gene Accession No. Q9BW85, Q9D6J3
Gene Alias 2900016D05Rik; AI413813; Ccdc94; coiled-coil domain containing 94; coiled-coil domain-containing protein 94; Splicing factor YJU2; YJU2
Gene Symbols CCDC94
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC94. Recombinant protein control fragment (Product #RP-98975).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55702, 72886
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.