Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC94 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen CCDC94
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126521
SDP
View Documents
Novus Biologicals
NBP310810100UL
100 μg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

CCDC94 Polyclonal specifically detects CCDC94 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CCDC94
Western Blot
Unconjugated
Rabbit
Human
coiled-coil domain containing 94, coiled-coil domain-containing protein 94, FLJ10374
The immunogen is a synthetic peptide directed towards the N terminal region of human CCDC94 (NP_060544.2). Peptide sequence MSERKVLNKYYPPDFDPSKIPKLKLPKDRQYVVRLMAPFNMRCKTCGEYI
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
55702
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.