Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC96 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561542 Supplier Invitrogen™ Supplier No. PA561542
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEELEAEAGA EELEQAAEGK EVRFQASLPL TRIDEEEAAA APEAETERVE GEEEDKEETQ RDGAESKERD GEGRPAKSQE EGKRLYGRDE F Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 33%.

CCDC96 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC96
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC96
Gene Accession No. Q2M329
Gene Alias 4921513E08Rik; CCDC96; coiled-coil domain containing 96; coiled-coil domain-containing protein 96; RGD1305153
Gene Symbols CCDC96
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC96. Recombinant protein control fragment (Product #RP-100234).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257236
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.