Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC96 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988311
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8311 Supplier Bioss Supplier No. BS8144R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC96 is a protein coding gene.

Specifications

Antigen CCDC96
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CCDC96
Gene Accession No. Q2M329
Gene Alias CCDC96; coiled-coil domain containing 96; Coiled-coil domain-containing protein 96
Gene Symbols CCDC96
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CCDC96.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257236
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.