Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC99 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CCDC99 |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CCDC99 Polyclonal specifically detects CCDC99 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
CCDC99 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
54908 | |
This antibody was developed against a recombinant protein corresponding to amino acids: YEPEETVEVPVLKKRREVLPVDITTAKDACVNNSALGGEVYRLPPQKEETQSCPNSLEDNNLQLEKSVSIHTPVVSLSPHKNLPVDMQLKKEKKCVKLIGVPADA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Human | |
Arsenite-related gene 1 protein, coiled-coil domain containing 99, Coiled-coil domain-containing protein 99, FLJ20364, FLJ40690, hSpindly, protein Spindly, Rhabdomyosarcoma antigen MU-RMS-40.4A, rrhabdomyosarcoma antigen protein MU-RMS-40.4A, Spindly | |
SPDL1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title