Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595815
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595815 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595815 Supplier Invitrogen™ Supplier No. PA595815
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTQTVNERED PLNLGGGGWA SSIPLRTWSS YHRRQRGAPV SKRRYRDGPK IEYEASRKQP KQQRSPGSWF QPSRGPYWAL YSNWERCGGP WRPPLIAFQS PLCPAQMIRA YGLHPLCVCC CSCWSGPWNP GWERPPGRKK RWGRRGRGLR RHPRRSFPRN PPIDLSKMLR PVNLSGWRAP GMRAPRNTTQ FIMNQVYEDM; Positive Samples: DU145.

CCER1 (Coiled-Coil Glutamate Rich Protein 1) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCER1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CCER1
Gene Accession No. Q6AY45, Q8TC90, Q9CQL2
Gene Alias C12orf12; CCER1; Coiled-coil domain-containing glutamate-rich protein 1; coiled-coil glutamate rich protein 1; coiled-coil glutamate-rich protein 1
Gene Symbols CCER1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of mouse Ccer1 (NP_0800001).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196477, 500825, 66716
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.