Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103116 Supplier Invitrogen™ Supplier No. PA5103116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cholecystokinin/CCK.

Cholecystokinin (CCK) is a neuropeptide hormone and neurotransmitter widely distributed throughout the gastrointestinal (GI) tract and the central nervous system (CNS). Antibodies that react specifically with CCK are useful for the study of the mode of action, differential tissue expression and intracellular and subcellular localization of CCK in the CNS and periphery as well as in neuroendocrine cells of the digestive system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CCK
Gene Accession No. P01355, P06307, P09240
Gene Alias (1-49)-CCK58; CCK; CCK12; CCK18; CCK22; CCK25; CCK33; CCK39; CCK5; CCK58; CCK7; CCK8; Cholecystokinin; cholecystokinin octapeptide; cholecystokinin triacontatriapeptide; Cholecystokinin-12; Cholecystokinin-18; Cholecystokinin-22; Cholecystokinin-25; Cholecystokinin-33; Cholecystokinin-39; Cholecystokinin-5; Cholecystokinin-58; Cholecystokinin-58 desnonopeptide; Cholecystokinin-7; Cholecystokinin-8; prepro-cholecystokinin
Gene Symbols CCK
Host Species Rabbit
Immunogen A synthesized peptide derived from human CCK(Accession P06307), corresponding to amino acid residues Q68-S115.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12424, 25298, 885
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.