Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584814
Description
Immunogen sequence: LQRAEEAPRR QLRVSQRTDG ESRAHLGALL ARYIQQARKA PSGRMSIVKN LQNLDPSHRI SDRDYMG.
Cholecystokinin (CCK) is a neuropeptide hormone and neurotransmitter widely distributed throughout the gastrointestinal (GI) tract and the central nervous system (CNS). Antibodies that react specifically with CCK are useful for the study of the mode of action, differential tissue expression and intracellular and subcellular localization of CCK in the CNS and periphery as well as in neuroendocrine cells of the digestive system.
Specifications
CCK | |
Polyclonal | |
Unconjugated | |
CCK | |
(1-49)-CCK58; CCK; CCK12; CCK18; CCK22; CCK25; CCK33; CCK39; CCK5; CCK58; CCK7; CCK8; Cholecystokinin; cholecystokinin octapeptide; cholecystokinin triacontatriapeptide; Cholecystokinin-12; Cholecystokinin-18; Cholecystokinin-22; Cholecystokinin-25; Cholecystokinin-33; Cholecystokinin-39; Cholecystokinin-5; Cholecystokinin-58; Cholecystokinin-58 desnonopeptide; Cholecystokinin-7; Cholecystokinin-8; prepro-cholecystokinin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
885 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P06307 | |
CCK | |
Recombinant protein corresponding to Human CCK. Recombinant protein control fragment (Product #RP-106081). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction