Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCKAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518609
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518609 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518609 Supplier Invitrogen™ Supplier No. PA518609
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rabbit based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

This gene encodes a G-protein coupled receptor that binds non-sulfated members of the cholecystokinin family of peptide hormones. This receptor is a major physiologic mediator of pancreatic enzyme secretion and smooth muscle contraction of the gallbladder and stomach. In the central and peripheral nervous system this receptor regulates satiety and the release of beta-endorphin and dopamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCKAR
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CCKAR
Gene Accession No. P30551
Gene Alias AW106902; CCK1R; CCK1-R; CCK-A; ccka receptor; CCK-A receptor; Cckar; CCK-AR; CCKRA; cholecystokinin A receptor; cholecystokinin receptor type A; cholecystokinin type-A receptor; cholecystokinin-1 receptor; HGNC:1570
Gene Symbols CCKAR
Host Species Goat
Immunogen Synthetic peptide sequence (KFDASQKKSAKEKR) corresponding to the internal amino acids of Cckar (aa 256-266).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24889
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.