Learn More
Invitrogen™ CCKAR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532693
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Dog, Panda (100%); Bovine, Elephant, Horse, Rabbit (94%); Bat, Opossum, Guinea pig (88%); Mouse, Rat, Hamster (81%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment with Proteinase K.
This gene encodes a G-protein coupled receptor that binds non-sulfated members of the cholecystokinin family of peptide hormones. This receptor is a major physiologic mediator of pancreatic enzyme secretion and smooth muscle contraction of the gallbladder and stomach. In the central and peripheral nervous system this receptor regulates satiety and the release of beta-endorphin and dopamine.
Specifications
CCKAR | |
Polyclonal | |
Unconjugated | |
CCKAR | |
AW106902; CCK1R; CCK1-R; CCK-A; ccka receptor; CCK-A receptor; Cckar; CCK-AR; CCKRA; cholecystokinin A receptor; cholecystokinin receptor type A; cholecystokinin type-A receptor; cholecystokinin-1 receptor; HGNC:1570 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
488846, 886 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P32238, Q5D0K2 | |
CCKAR | |
Synthetic 16 amino acid peptide from internal region of human CCKAR. | |
50 μg | |
Primary | |
Human, Canine, Monkey | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.