Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCKAR Polyclonal Antibody, Invitrogen™

Catalog No. PIPA131121
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA131121 50 μL
1 options

Catalog No. PIPA131121

Supplier: Thermo Scientific PA131121

Rabbit Polyclonal Antibody

PA1-31121 detects CCKAR from human samples.

This gene encodes a G-protein coupled receptor that binds non-sulfated members of the cholecystokinin family of peptide hormones. This receptor is a major physiologic mediator of pancreatic enzyme secretion and smooth muscle contraction of the gallbladder and stomach. In the central and peripheral nervous system this receptor regulates satiety and the release of beta-endorphin and dopamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCKAR
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with no preservative
Gene CCKAR
Gene Accession No. P32238
Gene Alias AW106902; CCK1R; CCK1-R; CCK-A; ccka receptor; CCK-A receptor; Cckar; CCK-AR; CCKRA; cholecystokinin A receptor; cholecystokinin receptor type A; cholecystokinin type-A receptor; cholecystokinin-1 receptor; HGNC:1570
Gene Symbols CCKAR
Host Species Rabbit
Immunogen Poly Lysin conjugated synthetic peptide: RFLLPPNDVMQQSWH, corresponding to amino acids 197-210 of Human CCKAR.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.